Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 9 - 10 | 1299 | 96 | |
| NM_172696.2 | NP_766284.2 | 9 - 10 | 1299 | 96 | ||
| GenBank mRNA | AK050545.1 | - | 9 - 10 | 1271 | 96 | |
| AK136048.1 | - | 3 - 4 | 482 | 96 | ||
| AK147866.1 | - | 9 - 10 | 1248 | 96 | ||
| BC050846.1 | - | 9 - 10 | 1233 | 96 | ||
| BC141404.1 | - | 9 - 10 | 1275 | 9056017 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inadl
Genomic Map