Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 10 - 11 | 1391 | 86 | |
| NM_172696.2 | NP_766284.2 | 10 - 11 | 1391 | 86 | ||
| GenBank mRNA | AK050545.1 | - | 10 - 11 | 1363 | 86 | |
| AK136048.1 | - | 4 - 5 | 574 | 86 | ||
| AK147866.1 | - | 10 - 11 | 1340 | 86 | ||
| BC050846.1 | - | 10 - 11 | 1325 | 86 | ||
| BC141404.1 | - | 10 - 11 | 1367 | 9056017 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inadl
Genomic Map