Assay Details
| Gene Symbol: Rbm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102407.1 | NP_001095877.1 | 3 - 4 | 237 | 62 | |
| NM_025875.2 | NP_080151.2 | 3 - 4 | 234 | 62 | ||
| GenBank mRNA | AK009953.1 | - | 3 - 4 | 229 | 62 | |
| AK010284.1 | - | 3 - 4 | 222 | 62 | ||
| AK082925.1 | - | 3 - 4 | 226 | 62 | ||
| BC020086.1 | - | 3 - 4 | 217 | 3671644 | 62 | |
| BC058376.1 | - | 3 - 4 | 206 | 5709938 | 62 | |
| BY128307.1 | - | 3 - 4 | 231 | 62 | ||
| KF051021.1 | - | 3 - 4 | 203 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm8a
Genomic Map