Assay Details
| Gene Symbol: Hfe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027126.4 | NP_081402.3 | 3 - 4 | 869 | 79 | |
| GenBank mRNA | AJ557515.1 | - | 3 - 4 | 869 | 79 | |
| AK014082.1 | - | 3 - 4 | 842 | 79 | ||
| BC022603.1 | - | 3 - 4 | 893 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014082.1, BC022603.1 transcripts.
Set Membership: |
|

Hfe2
Genomic Map