Assay Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178704.3 | NP_848819.2 | 12 - 13 | 1562 | 71 | |
| GenBank mRNA | AF538322.1 | - | 12 - 13 | 1546 | 71 | |
| AK033395.1 | - | 12 - 13 | 1519 | 71 | ||
| AK034217.1 | - | 12 - 13 | 1563 | 71 | ||
| AK045758.1 | - | 12 - 13 | 1463 | 71 | ||
| BC050065.1 | - | 12 - 13 | 1506 | 30016211 | 71 | |
| BC059897.1 | - | 12 - 13 | 1459 | 6821433 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l3
Genomic Map