Assay Details
Gene Symbol: Dpy19l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178704.3 | NP_848819.2 | 13 - 14 | 1687 | 97 | |
GenBank mRNA | AF538322.1 | - | 13 - 14 | 1671 | 97 | |
AK031345.1 | - | 13 - 14 | 1679 | 97 | ||
AK033395.1 | - | 13 - 14 | 1644 | 97 | ||
AK034217.1 | - | 13 - 14 | 1688 | 97 | ||
AK045758.1 | - | 13 - 14 | 1588 | 97 | ||
BC050065.1 | - | 13 - 14 | 1631 | 30016211 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
