Assay Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178704.3 | NP_848819.2 | 1 - 2 | 199 | 110 | |
| GenBank mRNA | AF538322.1 | - | 1 - 2 | 183 | 110 | |
| AK031345.1 | - | 1 - 2 | 191 | 110 | ||
| AK031722.1 | - | 1 - 2 | 101 | 110 | ||
| AK033395.1 | - | 1 - 2 | 156 | 110 | ||
| AK034217.1 | - | 1 - 2 | 200 | 110 | ||
| AK045758.1 | - | 1 - 2 | 100 | 110 | ||
| BC050065.1 | - | 1 - 2 | 143 | 30016211 | 110 | |
| BC059897.1 | - | 1 - 2 | 96 | 6821433 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF538322.1, AK033395.1, AK034217.1, AK045758.1, BC050065.1, BC059897.1, NM_178704.3 transcripts.
Set Membership: |
|

Dpy19l3
Genomic Map