Assay Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026360.3 | NP_080636.2 | 10 - 11 | 1139 | 63 | |
| GenBank mRNA | AK010310.1 | - | 10 - 11 | 1132 | 63 | |
| AK076982.1 | - | 10 - 11 | 1124 | 63 | ||
| AK169595.1 | - | 8 - 9 | 762 | 63 | ||
| BC006843.1 | - | 3 - 4 | 326 | 63 | ||
| BC095944.1 | - | 10 - 11 | 1116 | 4935970 | 63 | |
| BC132242.1 | - | 10 - 11 | 1131 | 40130519 | 63 | |
| BC132244.1 | - | 10 - 11 | 1131 | 40130521 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx47
Genomic Map