Assay Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026360.3 | NP_080636.2 | 5 - 6 | 594 | 95 | |
| GenBank mRNA | AK010310.1 | - | 5 - 6 | 587 | 95 | |
| AK076982.1 | - | 5 - 6 | 579 | 95 | ||
| AK169595.1 | - | 3 - 4 | 217 | 95 | ||
| BC060168.1 | - | 5 - 6 | 574 | 95 | ||
| BC095944.1 | - | 5 - 6 | 571 | 4935970 | 95 | |
| BC132242.1 | - | 5 - 6 | 586 | 40130519 | 95 | |
| BC132244.1 | - | 5 - 6 | 586 | 40130521 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx47
Genomic Map