Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024707.2 | NP_001019878.1 | 3 - 4 | 600 | 100 | |
| GenBank mRNA | AK149314.1 | - | 2 - 3 | 125 | 100 | |
| AK149337.1 | - | 596 | 100 | |||
| CJ106421.1 | - | 3 - 4 | 603 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp3
Genomic Map