Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024707.2 | NP_001019878.1 | 6 - 7 | 2065 | 71 | |
| GenBank mRNA | AK149314.1 | - | 5 - 6 | 1590 | 71 | |
| AK149337.1 | - | 2061 | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp3
Genomic Map