Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024707.2 | NP_001019878.1 | 7 - 8 | 2207 | 73 | |
| GenBank mRNA | AK149314.1 | - | 6 - 7 | 1732 | 73 | |
| BC094349.1 | - | 1 - 2 | 301 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024874 | BC094349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024874 | AK149314.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024874 | AK149337.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024874 | NM_001024707.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141924 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024874 | BC094349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024874 | AK149314.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024874 | AK149337.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024874 | NM_001024707.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06162604 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01496155 | - | - |
| Whole Mouse Genome G4122A | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P178866 | BC094349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P178866 | AK149314.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P178866 | NM_001024707.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06162604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25048290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06174929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06080259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06246768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06088698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06195384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06316111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06460845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06174924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06174930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25048259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562424 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436083_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562419 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552143 | - | - |
| Mouse Genome 430 2.0 Array | 1436083_at | - | - |
| Mouse Genome 430 2.0 Array | 1436083_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700025415.mm.1 | TC0700002846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025415.mm.1 | NM_001024707.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025417.mm.1 | TC0700002846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025417.mm.1 | NM_001024707.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025416.mm.1 | TC0700002846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025416.mm.1 | NM_001024707.2 | Refseq |
| Mouse Expression Set 430 | 1436083_at | - | - |
| Mouse Expression Set 430 | 1436083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436083_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 560696 | - | - |

Lrp3
Genomic Map