Assay Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172632.2 | NP_766220.2 | 2 - 3 | 1496 | 70 | |
| GenBank mRNA | AK079654.1 | - | 2 - 3 | 1474 | 70 | |
| BC058942.1 | - | 1 - 2 | 899 | 6821544 | 70 | |
| BC062911.1 | - | 1 - 2 | 899 | 6821544 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk4
Genomic Map