Assay Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172632.2 | NP_766220.2 | 4 - 5 | 1804 | 84 | |
| GenBank mRNA | AK079654.1 | - | 4 - 5 | 1782 | 84 | |
| BC058942.1 | - | 3 - 4 | 1207 | 6821544 | 84 | |
| BC062911.1 | - | 3 - 4 | 1207 | 6821544 | 84 | |
| BC094564.1 | - | 3 - 4 | 2018 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk4
Genomic Map