Assay Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008891.2 | NP_032917.2 | 7 - 8 | 741 | 135 | |
| GenBank mRNA | AK028219.1 | - | 7 - 8 | 682 | 135 | |
| AK030978.1 | - | 7 - 8 | 670 | 135 | ||
| AK050669.1 | - | 7 - 8 | 687 | 135 | ||
| AK085295.1 | - | 5 - 6 | 1846 | 135 | ||
| AK157587.1 | - | 7 - 8 | 685 | 135 | ||
| AK160616.1 | - | 7 - 8 | 676 | 135 | ||
| AK161109.1 | - | 7 - 8 | 681 | 135 | ||
| BC132571.1 | - | 7 - 8 | 675 | 40130848 | 135 | |
| BC138014.1 | - | 7 - 8 | 675 | 8861033 | 135 | |
| Y08701.1 | - | 7 - 8 | 672 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470080 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470083 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125831 | CJ318297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125831 | AK030978.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125831 | NM_008891.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64887 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136569 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125831 | CJ318297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125831 | AK030978.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125831 | NM_008891.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P121793 | Y08701.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470092 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136569 | - | - |
| Whole Mouse Genome G4122A | A_51_P121793 | Y08701.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106383 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125831 | CJ318297.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125831 | AK030978.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125831 | NM_008891.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136569 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09769820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470080 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470083 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28470084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09960077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09786508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09786740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10012856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09931870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09799807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470129 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395921 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423324_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450938_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423325_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395910 | - | - |
| Mouse Genome 430 2.0 Array | 1450938_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423324_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423325_at | - | - |
| Mouse Genome 430 2.0 Array | 1450938_at | - | - |
| Mouse Genome 430 2.0 Array | 1423325_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423324_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18949_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18949_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003477.mm.1 | NM_008891.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003477.mm.1 | NM_008891.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003477.mm.1 | TC1200000500.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003478.mm.1 | NM_008891.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003478.mm.1 | NM_008891.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003478.mm.1 | TC1200000500.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003475.mm.1 | NM_008891.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003475.mm.1 | NM_008891.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003475.mm.1 | TC1200000500.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003479.mm.1 | NM_008891.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003479.mm.1 | NM_008891.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003479.mm.1 | TC1200000500.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450938_PM_at | - | - |
| Mouse Expression Set 430 | 1450938_at | - | - |
| Mouse Expression Set 430 | 1423325_at | - | - |
| Mouse Expression Set 430 | 1423324_PM_at | - | - |
| Mouse Expression Set 430 | 1423324_at | - | - |
| Mouse Expression Set 430 | 1423325_PM_at | - | - |
| Murine Genome U74v2 Set | 94239_at | - | - |
| Murine Genome U74v2 Set | 94239_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450938_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423325_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423324_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423324_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423325_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450938_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450938_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423325_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423324_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423324_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423325_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450938_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 587998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6650504 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650504 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101980017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650504 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773034 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773034 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224089 | - | - |
| mouse-6 v1.1 (Illumina) | scl018949.11_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650504 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101980017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl018949.11_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101980017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650504 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2773034 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224089 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773034 | - | - |

Pnn
Genomic Map