Assay Details
| Gene Symbol: Cog5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163126.1 | NP_001156598.1 | 3 - 4 | 313 | 64 | |
| GenBank mRNA | AK030656.1 | - | 3 - 4 | 314 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cog5
Genomic Map