Assay Details
| Gene Symbol: Nadk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159637.1 | NP_001153109.1 | 10 - 11 | 1278 | 69 | |
| NM_138671.2 | NP_619612.2 | 10 - 11 | 1177 | 69 | ||
| GenBank mRNA | AK147917.1 | - | 10 - 11 | 1255 | 69 | |
| AK164862.1 | - | 8 - 9 | 1362 | 69 | ||
| AK165510.1 | - | 10 - 11 | 1271 | 69 | ||
| AK170095.1 | - | 10 - 11 | 1259 | 69 | ||
| AK170608.1 | - | 10 - 11 | 1273 | 69 | ||
| AK170960.1 | - | 10 - 11 | 1259 | 69 | ||
| BC004012.1 | - | 10 - 11 | 1165 | 3493623 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nadk
Genomic Map