Assay Details
| Gene Symbol: Nadk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159637.1 | NP_001153109.1 | 9 - 10 | 1115 | 87 | |
| NM_138671.2 | NP_619612.2 | 9 - 10 | 1014 | 87 | ||
| GenBank mRNA | AK147917.1 | - | 9 - 10 | 1092 | 87 | |
| AK164862.1 | - | 7 - 8 | 1199 | 87 | ||
| AK165510.1 | - | 9 - 10 | 1108 | 87 | ||
| AK170095.1 | - | 9 - 10 | 1096 | 87 | ||
| AK170608.1 | - | 9 - 10 | 1110 | 87 | ||
| AK170960.1 | - | 9 - 10 | 1096 | 87 | ||
| BC004012.1 | - | 9 - 10 | 1002 | 3493623 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nadk
Genomic Map