Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 12 - 13 | 1494 | 85 | |
| NM_008860.3 | NP_032886.2 | 15 - 16 | 1694 | 85 | ||
| GenBank mRNA | AB110830.1 | - | 12 - 13 | 1454 | 85 | |
| AK042562.1 | - | 12 - 13 | 1452 | 85 | ||
| AK049959.1 | - | 12 - 13 | 1478 | 85 | ||
| BC057694.1 | - | 7 - 8 | 661 | 85 | ||
| BC072590.1 | - | 12 - 13 | 1494 | 85 | ||
| BC139761.1 | - | 15 - 16 | 1511 | 40109623 | 85 | |
| M94632.1 | - | 15 - 16 | 1511 | 85 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map