Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 13 - 14 | 1585 | 91 | |
| NM_008860.3 | NP_032886.2 | 16 - 17 | 1785 | 91 | ||
| GenBank mRNA | AK042562.1 | - | 13 - 14 | 1543 | 91 | |
| AK049959.1 | - | 13 - 14 | 1569 | 91 | ||
| AK147300.1 | - | 13 - 14 | 1403 | 91 | ||
| BC057694.1 | - | 8 - 9 | 752 | 91 | ||
| BC072590.1 | - | 13 - 14 | 1585 | 91 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map