Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 3 - 4 | 556 | 86 | |
| NM_008860.3 | NP_032886.2 | 6 - 7 | 756 | 86 | ||
| GenBank mRNA | AB110830.1 | - | 3 - 4 | 516 | 86 | |
| AK042562.1 | - | 3 - 4 | 514 | 86 | ||
| AK049959.1 | - | 3 - 4 | 540 | 86 | ||
| AK147300.1 | - | 3 - 4 | 374 | 86 | ||
| BC072590.1 | - | 3 - 4 | 556 | 86 | ||
| BC139761.1 | - | 6 - 7 | 573 | 40109623 | 86 | |
| M94632.1 | - | 6 - 7 | 573 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB110830.1, AK049959.1, AK147300.1, NM_001039079.2 transcripts.
Set Membership: |
|

Prkcz
Genomic Map