Assay Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155349.1 | - | 2 - 3 | 543 | 75 | |
| AK220168.1 | - | 2 - 3 | 507 | 75 | ||
| BC051991.1 | - | 2 - 3 | 302 | 75 | ||
| BC088987.1 | - | 2 - 3 | 407 | 75 | ||
| BC138848.1 | - | 3 - 4 | 414 | 8861870 | 75 | |
| BC138849.1 | - | 3 - 4 | 414 | 8861871 | 75 | |
| BU702464.1 | - | 2 - 3 | 204 | 75 | ||
| U10115.1 | - | 2 - 3 | 500 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl1
Genomic Map