Assay Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155349.1 | - | 3 - 4 | 665 | 72 | |
| BC051991.1 | - | 3 - 4 | 424 | 72 | ||
| BC088987.1 | - | 3 - 4 | 529 | 72 | ||
| BC138848.1 | - | 4 - 5 | 536 | 8861870 | 72 | |
| BC138849.1 | - | 4 - 5 | 536 | 8861871 | 72 | |
| BU702464.1 | - | 3 - 4 | 326 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl1
Genomic Map