Assay Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155349.1 | - | 6 - 7 | 1010 | 53 | |
| AK220168.1 | - | 5 - 6 | 1198 | 53 | ||
| BC138848.1 | - | 7 - 8 | 881 | 8861870 | 53 | |
| BC138849.1 | - | 7 - 8 | 881 | 8861871 | 53 | |
| BU702464.1 | - | 6 - 7 | 671 | 53 | ||
| U10115.1 | - | 6 - 7 | 967 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl1
Genomic Map