Assay Details
| Gene Symbol: Ttll10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029264.2 | NP_083540.1 | 8 - 9 | 911 | 91 | |
| GenBank mRNA | AK014686.1 | - | 6 - 7 | 1208 | 91 | |
| AK133079.1 | - | 8 - 9 | 912 | 91 | ||
| AM690754.1 | - | 5 - 6 | 795 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll10
Genomic Map