Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 4 - 5 | 629 | 71 | |
| GenBank mRNA | AB041605.1 | - | 4 - 5 | 606 | 71 | |
| AF211859.1 | - | 4 - 5 | 590 | 71 | ||
| AK007552.1 | - | 4 - 5 | 626 | 71 | ||
| AK008585.1 | - | 4 - 5 | 418 | 71 | ||
| AK044204.1 | - | 4 - 5 | 566 | 71 | ||
| AK048244.1 | - | 4 - 5 | 619 | 71 | ||
| AK156037.1 | - | 4 - 5 | 612 | 71 | ||
| AK169857.1 | - | 4 - 5 | 605 | 71 | ||
| AY337018.1 | - | 4 - 5 | 612 | 71 | ||
| BC023747.1 | - | 4 - 5 | 604 | 71 | ||
| BC132295.1 | - | 4 - 5 | 621 | 40130572 | 71 | |
| CD348864.1 | - | 4 - 5 | 629 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl2
Genomic Map