Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 8 - 9 | 1042 | 78 | |
| GenBank mRNA | AB041605.1 | - | 8 - 9 | 1019 | 78 | |
| AK048244.1 | - | 8 - 9 | 2822 | 78 | ||
| AY337018.1 | - | 8 - 9 | 1025 | 78 | ||
| BC132295.1 | - | 8 - 9 | 1034 | 40130572 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl2
Genomic Map