Assay Details
| Gene Symbol: Mmp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011985.2 | NP_036115.1 | 4 - 5 | 808 | 82 | |
| GenBank mRNA | AF085742.1 | - | 4 - 5 | 808 | 82 | |
| AK146510.1 | - | 4 - 5 | 615 | 82 | ||
| BC107357.1 | - | 4 - 5 | 715 | 40055939 | 82 | |
| BC107358.1 | - | 4 - 5 | 673 | 40055946 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp23
Genomic Map