Assay Details
| Gene Symbol: Mmp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011985.2 | NP_036115.1 | 7 - 8 | 1207 | 85 | |
| GenBank mRNA | AF085742.1 | - | 7 - 8 | 1207 | 85 | |
| AK146510.1 | - | 7 - 8 | 1014 | 85 | ||
| BC107357.1 | - | 7 - 8 | 1114 | 40055939 | 85 | |
| BC107358.1 | - | 7 - 8 | 1072 | 40055946 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439845 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680818 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137854 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23439825 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137854 | - | - |
| Whole Mouse Genome G4122A | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | BC107357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | BC107358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | NM_011985.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | AF085742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502132 | AK146510.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140150 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137854 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056497 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056557 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04056504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23439841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23439833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04056537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04056554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04056497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04056525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03254382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23439819 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10519148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519142 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519145 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417282_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417281_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10519140 | - | - |
| Mouse Genome 430 2.0 Array | 1417281_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417282_at | - | - |
| Mouse Genome 430 2.0 Array | 1417282_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417281_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26561_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26561_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400034742.mm.1 | TC0400004146.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400034742.mm.1 | NM_011985.3 | Refseq |
| Mouse Expression Set 430 | 1417281_PM_a_at | - | - |
| Mouse Expression Set 430 | 1417282_at | - | - |
| Mouse Expression Set 430 | 1417281_a_at | - | - |
| Mouse Expression Set 430 | 1417282_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417282_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417281_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417282_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417281_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417282_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417282_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417281_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417281_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 623672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3940278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237364 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237364 | - | - |
| mouse-6 v1.1 (Illumina) | scl23434.8.1_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23434.8.1_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237364 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237364 | - | - |

Mmp23
Genomic Map