Assay Details
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256107.1 | NP_001243036.1 | 14 - 15 | 2686 | 62 | |
NM_001256108.2 | NP_001243037.2 | 14 - 15 | 2506 | 62 | ||
NM_145124.3 | NP_660106.2 | 13 - 14 | 2590 | 62 | ||
GenBank mRNA | AK008671.1 | - | 1 - 2 | 80 | 62 | |
AK084791.1 | - | 6 - 7 | 888 | 62 | ||
AK147940.1 | - | 14 - 15 | 2451 | 62 | ||
AK159611.1 | - | 14 - 15 | 2403 | 62 | ||
AY974090.1 | - | 12 - 13 | 1887 | 62 | ||
BC058086.1 | - | 11 - 12 | 2174 | 62 | ||
BC141133.1 | - | 11 - 12 | 1979 | 9055745 | 62 | |
BC145441.1 | - | 12 - 13 | 2075 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
