Assay Details
| Gene Symbol: Mib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256107.1 | NP_001243036.1 | 15 - 16 | 2815 | 54 | |
| NM_001256108.2 | NP_001243037.2 | 15 - 16 | 2635 | 54 | ||
| NM_145124.3 | NP_660106.2 | 14 - 15 | 2719 | 54 | ||
| GenBank mRNA | AB072336.1 | - | 14 - 15 | 2281 | 54 | |
| AK008671.1 | - | 2 - 3 | 209 | 54 | ||
| AK084791.1 | - | 7 - 8 | 1017 | 54 | ||
| AK147940.1 | - | 15 - 16 | 2580 | 54 | ||
| AK159611.1 | - | 15 - 16 | 2532 | 54 | ||
| AY974090.1 | - | 13 - 14 | 2016 | 54 | ||
| BC058086.1 | - | 12 - 13 | 2303 | 54 | ||
| BC141133.1 | - | 12 - 13 | 2108 | 9055745 | 54 | |
| BC145441.1 | - | 13 - 14 | 2204 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mib2
Genomic Map