Assay Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007633.2 | NP_031659.2 | 5 - 6 | 521 | 99 | |
| GenBank mRNA | AK089950.1 | - | 5 - 6 | 453 | 99 | |
| AK163046.1 | - | 5 - 6 | 510 | 99 | ||
| AK165658.1 | - | 5 - 6 | 511 | 99 | ||
| BC003499.1 | - | 1 - 2 | 73 | 99 | ||
| BC062152.1 | - | 5 - 6 | 548 | 6510163 | 99 | |
| BC084588.1 | - | 5 - 6 | 565 | 6506897 | 99 | |
| BC106191.1 | - | 5 - 6 | 521 | 6336006 | 99 | |
| BC138660.1 | - | 4 - 5 | 354 | 8861680 | 99 | |
| BC138662.1 | - | 4 - 5 | 354 | 8861683 | 99 | |
| X75888.1 | - | 4 - 5 | 361 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccne1
Genomic Map