Assay Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007633.2 | NP_031659.2 | 9 - 10 | 1030 | 74 | |
| GenBank mRNA | AK089950.1 | - | 9 - 10 | 962 | 74 | |
| AK163046.1 | - | 9 - 10 | 1019 | 74 | ||
| AK165658.1 | - | 9 - 10 | 1020 | 74 | ||
| BC003499.1 | - | 5 - 6 | 582 | 74 | ||
| BC062152.1 | - | 9 - 10 | 1057 | 6510163 | 74 | |
| BC084588.1 | - | 9 - 10 | 1074 | 6506897 | 74 | |
| BC106191.1 | - | 9 - 10 | 1030 | 6336006 | 74 | |
| BC138660.1 | - | 8 - 9 | 863 | 8861680 | 74 | |
| BC138662.1 | - | 8 - 9 | 863 | 8861683 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccne1
Genomic Map