Assay Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007633.2 | NP_031659.2 | 10 - 11 | 1139 | 99 | |
| GenBank mRNA | AK089950.1 | - | 10 - 11 | 1071 | 99 | |
| AK163046.1 | - | 10 - 11 | 1128 | 99 | ||
| AK165658.1 | - | 10 - 11 | 1129 | 99 | ||
| BC003499.1 | - | 6 - 7 | 691 | 99 | ||
| BC062152.1 | - | 10 - 11 | 1166 | 6510163 | 99 | |
| BC084588.1 | - | 10 - 11 | 1183 | 6506897 | 99 | |
| BC106191.1 | - | 10 - 11 | 1139 | 6336006 | 99 | |
| BC138660.1 | - | 9 - 10 | 972 | 8861680 | 99 | |
| BC138662.1 | - | 9 - 10 | 972 | 8861683 | 99 | |
| X75888.1 | - | 9 - 10 | 979 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccne1
Genomic Map