Assay Details
| Gene Symbol: Pop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025390.4 | NP_079666.1 | 1 - 2 | 204 | 92 | |
| GenBank mRNA | AK002855.1 | - | 1 - 2 | 205 | 92 | |
| AK003908.1 | - | 1 - 2 | 219 | 92 | ||
| AK054466.1 | - | 236 | 92 | |||
| AK150459.1 | - | 1 - 2 | 211 | 92 | ||
| AK152948.1 | - | 1 - 2 | 211 | 92 | ||
| BC011465.1 | - | 1 - 2 | 220 | 3966371 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002855.1, AK150459.1, AK152948.1, BC011465.1, NM_025390.4 transcripts.
Set Membership: |
|

Pop4
Genomic Map