Assay Details
| Gene Symbol: Tmed7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025698.1 | NP_079974.1 | 1 - 2 | 445 | 77 | |
| GenBank mRNA | AK013936.1 | - | 1 - 2 | 428 | 77 | |
| AK029576.1 | - | 1 - 2 | 223 | 77 | ||
| AK147412.1 | - | 1 - 2 | 425 | 77 | ||
| AK202734.1 | - | 1 - 2 | 94 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmed7
Genomic Map