Assay Details
| Gene Symbol: Tmed7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025698.1 | NP_079974.1 | 2 - 3 | 692 | 64 | |
| GenBank mRNA | AK013936.1 | - | 2 - 3 | 675 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37579 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177062 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177062 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08988767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03467107 | - | - |
| Whole Mouse Genome G4122A | A_51_P269997 | BC024647.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269997 | AK077807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269997 | AV225029.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269997 | BC018585.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269997 | NM_025698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | CD565393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | AK147412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | AK076210.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500507 | NM_025698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121045 | AK147412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121045 | AK029576.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1419918_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419917_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427314_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429187_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427315_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458823 | - | - |
| Mouse Genome 430 2.0 Array | 1427315_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419918_at | - | - |
| Mouse Genome 430 2.0 Array | 1419918_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419917_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427314_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419917_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429187_at | - | - |
| Mouse Genome 430 2.0 Array | 1427314_at | - | - |
| Mouse Genome 430 2.0 Array | 1427315_at | - | - |
| Mouse Genome 430 2.0 Array | 1429187_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800009663.mm.1 | NM_025698.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009663.mm.1 | NM_025698.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009663.mm.1 | TC1800001752.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800012328.mm.1 | NM_025698.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012328.mm.1 | NM_025698.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012328.mm.1 | TC1800001752.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800009662.mm.1 | NM_025698.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009662.mm.1 | NM_025698.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009662.mm.1 | TC1800001752.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427315_PM_at | - | - |
| Mouse Expression Set 430 | 1427314_PM_at | - | - |
| Mouse Expression Set 430 | 1427314_at | - | - |
| Mouse Expression Set 430 | 1427315_at | - | - |
| Mouse Expression Set 430 | 1429187_PM_at | - | - |
| Mouse Expression Set 430 | 1429187_at | - | - |
| Murine Genome U74v2 Set | 94903_at | - | - |
| Murine Genome U74v2 Set | 160183_PM_f_at | - | - |
| Murine Genome U74v2 Set | 160183_f_at | - | - |
| Murine Genome U74v2 Set | 94903_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427315_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427315_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427314_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427314_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419917_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419918_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427314_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419917_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427314_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427315_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419918_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427315_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105340026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105340026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101780181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2579611 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2494869 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234851 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101780181 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105340026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105340026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101780181 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234851 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2579611 | - | - |

Tmed7
Genomic Map