Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112744.1 | NP_001106215.1 | 8 - 9 | 1400 | 81 | |
| GenBank mRNA | AK042195.1 | - | 8 - 9 | 1396 | 81 | |
| AK153708.1 | - | 8 - 9 | 1561 | 81 | ||
| BC020030.1 | - | 5 - 6 | 561 | 81 | ||
| BC095966.1 | - | 6 - 7 | 697 | 81 | ||
| BC137847.1 | - | 8 - 9 | 1441 | 8860866 | 81 | |
| BC144970.1 | - | 8 - 9 | 1441 | 9053518 | 81 | |
| BC144971.1 | - | 8 - 9 | 1464 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map