Assay Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113360.2 | NP_001106831.1 | 15 - 16 | 2362 | 56 | |
| NM_175556.4 | NP_780765.2 | 15 - 16 | 2362 | 56 | ||
| GenBank mRNA | AK129147.1 | - | 15 - 16 | 2122 | 56 | |
| AY966876.1 | - | 15 - 16 | 2140 | 56 | ||
| BC040465.1 | - | 3 - 4 | 373 | 56 | ||
| BC052329.1 | - | 5 - 6 | 495 | 56 | ||
| DQ176851.1 | - | 15 - 16 | 2362 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plch2
Genomic Map