Assay Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113360.2 | NP_001106831.1 | 8 - 9 | 1457 | 103 | |
| NM_175556.4 | NP_780765.2 | 8 - 9 | 1457 | 103 | ||
| GenBank mRNA | AK044619.1 | - | 8 - 9 | 1456 | 103 | |
| AK049970.1 | - | 8 - 9 | 1301 | 103 | ||
| AK078731.1 | - | 8 - 9 | 1228 | 103 | ||
| AK129147.1 | - | 8 - 9 | 1217 | 103 | ||
| AY966876.1 | - | 8 - 9 | 1235 | 103 | ||
| DQ176851.1 | - | 8 - 9 | 1457 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plch2
Genomic Map