Assay Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113360.2 | NP_001106831.1 | 9 - 10 | 1623 | 75 | |
| NM_175556.4 | NP_780765.2 | 9 - 10 | 1623 | 75 | ||
| GenBank mRNA | AK044619.1 | - | 9 - 10 | 1622 | 75 | |
| AK049970.1 | - | 9 - 10 | 1467 | 75 | ||
| AK078731.1 | - | 9 - 10 | 1394 | 75 | ||
| AK129147.1 | - | 9 - 10 | 1383 | 75 | ||
| AY966876.1 | - | 9 - 10 | 1401 | 75 | ||
| DQ176851.1 | - | 9 - 10 | 1623 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plch2
Genomic Map