Assay Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113360.2 | NP_001106831.1 | 10 - 11 | 1742 | 76 | |
| NM_175556.4 | NP_780765.2 | 10 - 11 | 1742 | 76 | ||
| GenBank mRNA | AK044619.1 | - | 10 - 11 | 1741 | 76 | |
| AK049970.1 | - | 10 - 11 | 1586 | 76 | ||
| AK129147.1 | - | 10 - 11 | 1502 | 76 | ||
| AY966876.1 | - | 10 - 11 | 1520 | 76 | ||
| DQ176851.1 | - | 10 - 11 | 1742 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plch2
Genomic Map