Assay Details
| Gene Symbol: Mylk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297612.1 | NP_001284541.1 | 7 - 8 | 1400 | 70 | |
| NM_175441.5 | NP_780650.2 | 7 - 8 | 1853 | 70 | ||
| GenBank mRNA | AK052858.1 | - | 7 - 8 | 1688 | 70 | |
| AK146683.1 | - | 7 - 8 | 1854 | 70 | ||
| AK165216.1 | - | 7 - 8 | 1342 | 70 | ||
| EU403565.1 | - | 7 - 8 | 1796 | 70 | ||
More Information
Set Membership: |
|

Mylk3
Genomic Map