Assay Details
| Gene Symbol: Mylk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297612.1 | NP_001284541.1 | 8 - 9 | 1537 | 84 | |
| NM_175441.5 | NP_780650.2 | 8 - 9 | 1990 | 84 | ||
| GenBank mRNA | AK052858.1 | - | 8 - 9 | 1825 | 84 | |
| AK146683.1 | - | 8 - 9 | 1991 | 84 | ||
| AK165216.1 | - | 8 - 9 | 1479 | 84 | ||
| BC137983.1 | - | 8 - 9 | 1963 | 8861002 | 84 | |
| BC145079.1 | - | 8 - 9 | 1964 | 84 | ||
| EU403565.1 | - | 8 - 9 | 1933 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mylk3
Genomic Map