Assay Details
| Gene Symbol: Spink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009258.5 | NP_033284.1 | 3 - 4 | 285 | 72 | |
| GenBank mRNA | AK007985.1 | - | 3 - 4 | 244 | 72 | |
| BC086887.1 | - | 3 - 4 | 244 | 6775994 | 72 | |
| CA481615.1 | - | 3 - 4 | 322 | 72 | ||
| X06342.1 | - | 3 - 4 | 242 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of peptidase activity
negative regulation of nitric oxide mediated signal transduction
negative regulation of peptidyl-tyrosine phosphorylation
regulation of acrosome reaction
negative regulation of calcium ion import
negative regulation of serine-type endopeptidase activity
regulation of store-operated calcium entry
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
negative regulation of peptidase activity
negative regulation of nitric oxide mediated signal transduction
negative regulation of peptidyl-tyrosine phosphorylation
regulation of acrosome reaction
negative regulation of calcium ion import
negative regulation of serine-type endopeptidase activity
regulation of store-operated calcium entry
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P365516 | X06342.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P365516 | X06342.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51503 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104760 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147446 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131268 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P365516 | X06342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P365516 | X06342.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104760 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131268 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147446 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134163 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122786 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147446 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104760 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13658854 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13658861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13658856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13658857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13858349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13738077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13794067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14041024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13658854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13658850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31663327 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10458705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458708 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415938_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458704 | - | - |
| Mouse Genome 430 2.0 Array | 1415938_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415938_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20730_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20730_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800009496.mm.1 | NM_009258.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009496.mm.1 | NM_009258.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009496.mm.1 | TC1800001274.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415938_at | - | - |
| Mouse Expression Set 430 | 1415938_PM_at | - | - |
| Murine Genome U74v2 Set | 96630_PM_at | - | - |
| Murine Genome U74v2 Set | 96630_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415938_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415938_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415938_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415938_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 348840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1580008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1580008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708478 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708478 | - | - |
| mouse-6 v1.1 (Illumina) | scl51454.4.1_44-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51454.4.1_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708477 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708477 | - | - |

Spink1
Genomic Map