Assay Details
Gene Symbol: Gm94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033280.2 | NP_001028452.1 | 3 - 4 | 323 | 64 | |
GenBank mRNA | AK131907.1 | - | 3 - 4 | 324 | 64 | |
AK131908.1 | - | 3 - 4 | 324 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | AK131907.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | AK131908.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | AK131907.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P351378 | AK131908.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331607 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187747 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905762 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176732 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132546 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | AK131907.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | AK131908.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | AK131907.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P351378 | AK131908.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663663 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31663635 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03462834 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132546 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08982857 | - | - |
Whole Mouse Genome G4122A | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P351378 | AK131907.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P351378 | AK131908.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P351378 | NM_001033280.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P351378 | AK131907.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P351378 | AK131908.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31663523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14056158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14130581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13812924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663680 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663575 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14056153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663597 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663580 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663625 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13625234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13812933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14056165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13939609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663626 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13924598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31663659 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10458710 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10458713 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10458712 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10458711 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1458367_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10458709 | - | - |
Mouse Genome 430 2.0 Array | 1458367_at | - | - |
Mouse Genome 430 2.0 Array | 1458367_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225443_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225443_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800009501.mm.1 | NM_001033280.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800009501.mm.1 | NM_001033280.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800009501.mm.1 | TC1800001275.mm.1 | MTA TC |
Mouse Expression Set 430 | 1458367_at | - | - |
Mouse Expression Set 430 | 1458367_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458367_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458367_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 801975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243054 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2900502 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2900502 | - | - |
