Assay Details
| Gene Symbol: Csf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037859.2 | NP_001032948.2 | 16 - 17 | 2502 | 120 | |
| GenBank mRNA | AK004947.1 | - | 15 - 16 | 2312 | 120 | |
| AK076215.1 | - | 15 - 16 | 2311 | 120 | ||
| AK079247.1 | - | 15 - 16 | 2319 | 120 | ||
| AK080326.1 | - | 15 - 16 | 2300 | 120 | ||
| AK143545.1 | - | 15 - 16 | 2303 | 120 | ||
| AK146069.1 | - | 16 - 17 | 2503 | 120 | ||
| AK154128.1 | - | 15 - 16 | 2317 | 120 | ||
| AK154556.1 | - | 15 - 16 | 2313 | 120 | ||
| AK154653.1 | - | 15 - 16 | 2313 | 120 | ||
| AK154699.1 | - | 15 - 16 | 2313 | 120 | ||
| AK155488.1 | - | 15 - 16 | 2323 | 120 | ||
| AK155498.1 | - | 1 - 2 | 79 | 120 | ||
| AK155576.1 | - | 2319 | 120 | |||
| AK155640.1 | - | 15 - 16 | 2317 | 120 | ||
| AK155654.1 | - | 15 - 16 | 2317 | 120 | ||
| AK155858.1 | - | 15 - 16 | 2320 | 120 | ||
| AK156550.1 | - | 15 - 16 | 2337 | 120 | ||
| AK171386.1 | - | 15 - 16 | 2317 | 120 | ||
| BC036343.1 | - | 15 - 16 | 2289 | 4948135 | 120 | |
| BC043054.1 | - | 15 - 16 | 2293 | 6418718 | 120 | |
| BC050024.1 | - | 15 - 16 | 2400 | 120 | ||
| BC066863.1 | - | 16 - 17 | 2411 | 30436119 | 120 | |
| X06368.1 | - | 15 - 16 | 120 | |||
| X68932.1 | - | 15 - 16 | 120 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Csf1r
Genomic Map