Assay Details
| Gene Symbol: Csf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037859.2 | NP_001032948.2 | 18 - 19 | 2720 | 92 | |
| GenBank mRNA | AK004947.1 | - | 17 - 18 | 2530 | 92 | |
| AK076215.1 | - | 17 - 18 | 2529 | 92 | ||
| AK079247.1 | - | 17 - 18 | 2537 | 92 | ||
| AK080326.1 | - | 17 - 18 | 2518 | 92 | ||
| AK143545.1 | - | 17 - 18 | 2521 | 92 | ||
| AK146069.1 | - | 18 - 19 | 2721 | 92 | ||
| AK154128.1 | - | 17 - 18 | 2535 | 92 | ||
| AK154556.1 | - | 17 - 18 | 2531 | 92 | ||
| AK154653.1 | - | 17 - 18 | 2531 | 92 | ||
| AK154699.1 | - | 17 - 18 | 2531 | 92 | ||
| AK155488.1 | - | 17 - 18 | 2541 | 92 | ||
| AK155498.1 | - | 3 - 4 | 297 | 92 | ||
| AK155565.1 | - | 13 - 14 | 2030 | 92 | ||
| AK155576.1 | - | 2537 | 92 | |||
| AK155640.1 | - | 17 - 18 | 2535 | 92 | ||
| AK155654.1 | - | 17 - 18 | 2535 | 92 | ||
| AK155858.1 | - | 17 - 18 | 2538 | 92 | ||
| AK156550.1 | - | 17 - 18 | 2555 | 92 | ||
| AK171386.1 | - | 17 - 18 | 2535 | 92 | ||
| BC036343.1 | - | 17 - 18 | 2507 | 4948135 | 92 | |
| BC043054.1 | - | 17 - 18 | 2511 | 6418718 | 92 | |
| BC050024.1 | - | 17 - 18 | 2618 | 92 | ||
| BC066863.1 | - | 18 - 19 | 2629 | 30436119 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csf1r
Genomic Map