Assay Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028127.3 | NP_082403.2 | 1 - 2 | 66 | 90 | |
| GenBank mRNA | BC053929.1 | - | 1 - 2 | 66 | 6389695 | 90 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC053929.1, NM_028127.3 transcripts.
Set Membership: |
|

Frmd6
Genomic Map