Assay Details
| Gene Symbol: Sap30bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020483.3 | NP_065229.2 | 1 - 2 | 118 | 61 | |
| GenBank mRNA | AK147869.1 | - | 1 - 2 | 140 | 61 | |
| BC021757.1 | - | 1 - 2 | 118 | 5065414 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sap30bp
Genomic Map